Assay Details
| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011145.3 | NP_035275.1 | 3 - 4 | 401 | 72 | |
| GenBank mRNA | AB221579.1 | - | 1 - 2 | 130 | 72 | |
| AK089913.1 | - | 3 - 4 | 349 | 72 | ||
| AK154567.1 | - | 3 - 4 | 391 | 72 | ||
| BC070398.1 | - | 3 - 4 | 352 | 5694282 | 72 | |
| CA978861.1 | - | 3 - 4 | 401 | 72 | ||
| L28116.1 | - | 1 - 2 | 183 | 72 | ||
| U01665.1 | - | 1 - 2 | 194 | 72 | ||
| U10375.1 | - | 1 - 2 | 126 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppard
Genomic Map