Assay Details
| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011145.3 | NP_035275.1 | 4 - 5 | 555 | 79 | |
| GenBank mRNA | AB221579.1 | - | 2 - 3 | 284 | 79 | |
| AK089913.1 | - | 4 - 5 | 503 | 79 | ||
| BC070398.1 | - | 4 - 5 | 506 | 5694282 | 79 | |
| CA978861.1 | - | 4 - 5 | 555 | 79 | ||
| L28116.1 | - | 2 - 3 | 337 | 79 | ||
| U01665.1 | - | 2 - 3 | 348 | 79 | ||
| U10375.1 | - | 2 - 3 | 280 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppard
Genomic Map