Assay Details
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010269.3 | NP_034399.1 | 8 - 9 | 1224 | 66 | |
| NR_104341.1 | 8 - 9 | 1186 | 66 | |||
| GenBank mRNA | AK004887.1 | - | 8 - 9 | 1170 | 66 | |
| AK082958.1 | - | 8 - 9 | 1186 | 66 | ||
| AK146835.1 | - | 8 - 9 | 1214 | 66 | ||
| AK161122.1 | - | 8 - 9 | 1123 | 66 | ||
| AK166799.1 | - | 8 - 9 | 1139 | 66 | ||
| AK166859.1 | - | 8 - 9 | 1138 | 66 | ||
| BC025070.1 | - | 8 - 9 | 1133 | 3710712 | 66 | |
| Y17851.1 | - | 8 - 9 | 1221 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdap2
Genomic Map