Assay Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028015.2 | NP_082291.1 | 6 - 7 | 778 | 111 | |
| GenBank mRNA | AK010241.1 | - | 6 - 7 | 706 | 111 | |
| AK075694.1 | - | 6 - 7 | 698 | 111 | ||
| AK145589.1 | - | 6 - 7 | 686 | 111 | ||
| AK145762.1 | - | 6 - 7 | 700 | 111 | ||
| AK146417.1 | - | 6 - 7 | 669 | 111 | ||
| AY029533.1 | - | 6 - 7 | 698 | 111 | ||
| BC043059.1 | - | 725 | 6419704 | 111 | ||
| BC046797.1 | - | 6 - 7 | 778 | 6403129 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers5
Genomic Map