Assay Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028015.2 | NP_082291.1 | 8 - 9 | 1008 | 104 | |
| GenBank mRNA | AK010241.1 | - | 8 - 9 | 936 | 104 | |
| AK075694.1 | - | 8 - 9 | 928 | 104 | ||
| AK145589.1 | - | 8 - 9 | 916 | 104 | ||
| AK145762.1 | - | 8 - 9 | 930 | 104 | ||
| AK146220.1 | - | 8 - 9 | 930 | 104 | ||
| AK146417.1 | - | 8 - 9 | 900 | 104 | ||
| AY029533.1 | - | 8 - 9 | 928 | 104 | ||
| BC010670.1 | - | 2 - 3 | 229 | 104 | ||
| BC043059.1 | - | 955 | 6419704 | 104 | ||
| BC046797.1 | - | 8 - 9 | 1008 | 6403129 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers5
Genomic Map