Assay Details
| Gene Symbol: Spag17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028892.4 | NP_083168.3 | 12 - 13 | 1690 | 72 | |
| GenBank mRNA | AK016477.1 | - | 12 - 13 | 1676 | 72 | |
| AK029691.1 | - | 12 - 13 | 1691 | 72 | ||
| AK029947.1 | - | 12 - 13 | 1686 | 72 | ||
| AY555275.1 | - | 12 - 13 | 1676 | 72 | ||
| AY792594.1 | - | 12 - 13 | 1676 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spag17
Genomic Map