Assay Details
| Gene Symbol: Gpatch2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027405.2 | NP_081681.2 | 1 - 2 | 66 | 61 | |
| GenBank mRNA | AK031712.1 | - | 1 - 2 | 70 | 61 | |
| AK129299.1 | - | 1 - 2 | 43 | 61 | ||
| AK147916.1 | - | 1 - 2 | 55 | 61 | ||
| BC048169.1 | - | 1 - 2 | 75 | 6821902 | 61 | |
| BC050782.1 | - | 1 - 2 | 46 | 61 | ||
| BC058256.1 | - | 1 - 2 | 66 | 6466603 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031712.1, AK147916.1, BC048169.1, BC058256.1, NM_027405.2 transcripts.
Set Membership: |
|

Gpatch2l
Genomic Map