| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
NR_045065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000795.mm.1 |
TC1500000084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
NR_045065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000797.mm.1 |
TC1500000084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000803.mm.1 |
NR_045065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000803.mm.1 |
NR_045065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000803.mm.1 |
TC1500000084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
NR_045065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000794.mm.1 |
TC1500000084.mm.1 |
MTA TC |