Assay Details
| Gene Symbol: Car10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038695.1 | - | 1 - 2 | 229 | 59 | |
| BC066111.1 | - | 1 - 2 | 305 | 30360743 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066111.1 transcripts.
Set Membership: |
|

Car10
Genomic Map