Assay Details
| Gene Symbol: Galnt6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028111.1 | - | 437 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 | Galnt6 |
4632410F13, AW047994, GalNAc-T6 |
Anti-sense | Chr. 15 - 100689915 - 100729929 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA I730030J21 gene | I730030J21Rik | 26.9kb | Anti-sense | Chr. 15 - 100730505 - 100732737 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Galnt6os
Genomic Map