Assay Details
| Gene Symbol: Gm15471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040412.1 | 4 - 5 | 402 | 90 | ||
| GenBank mRNA | AK052792.1 | - | 3 - 4 | 1008 | 90 | |
| AW823483.1 | - | 1 - 2 | 90 | |||
| BX516086.1 | - | 4 - 5 | 368 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeodomain interacting protein kinase 1 | Hipk1 |
1110062K04Rik, Myak |
-13.0kb | Anti-sense | Chr. 3 - 103739815 - 103791894 on NCBI Build GRCm38 |
| Downstream |
adaptor-related protein complex AP-4, beta 1 |
Ap4b1 |
1810038H16Rik, AV004952, Ap4b4 |
1.2kb | Sense | Chr. 3 - 103809517 - 103822025 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15471
Genomic Map