Assay Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019505.3 | NP_062378.1 | 2 - 3 | 977 | 95 | |
| GenBank mRNA | AF136744.1 | - | 2 - 3 | 614 | 95 | |
| AK044596.1 | - | 1713 | 95 | |||
| AK049335.1 | - | 2 - 3 | 777 | 95 | ||
| AK147841.1 | - | 2 - 3 | 978 | 95 | ||
| BB635763.1 | - | 2 - 3 | 381 | 95 | ||
| BC055867.1 | - | 2 - 3 | 910 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgke
Genomic Map