Assay Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019505.3 | NP_062378.1 | 8 - 9 | 1641 | 155 | |
| GenBank mRNA | AF136744.1 | - | 8 - 9 | 1278 | 155 | |
| AK049335.1 | - | 8 - 9 | 1441 | 155 | ||
| BC055867.1 | - | 8 - 9 | 1573 | 155 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgke
Genomic Map