Assay Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019505.3 | NP_062378.1 | 9 - 10 | 1764 | 67 | |
| GenBank mRNA | AF136744.1 | - | 9 - 10 | 1401 | 67 | |
| AK049335.1 | - | 9 - 10 | 1564 | 67 | ||
| AK147841.1 | - | 9 - 10 | 1801 | 67 | ||
| BC055867.1 | - | 9 - 10 | 1696 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dgke
Genomic Map