Assay Details
| Gene Symbol: 1810013A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045427.1 | 2 - 3 | 226 | 65 | ||
| GenBank mRNA | AK007468.1 | - | 2 - 3 | 227 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | differentially expressed in FDCP 6 | Def6 |
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6 |
-43.6kb | Sense | Chr. 17 - 28207778 - 28228608 on NCBI Build GRCm38 |
| Downstream |
Fanconi anemia, complementation group E |
Fance |
2810451D06Rik, AI415634, AW209126 |
40.4kb | Sense | Chr. 17 - 28313530 - 28326574 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1810013A23Rik
Genomic Map