Assay Details
| Gene Symbol: Gm15886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016162.1 | - | 2 - 3 | 125 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeodomain interacting protein kinase 1 | Hipk1 |
1110062K04Rik, Myak |
Anti-sense | Chr. 3 - 103739815 - 103791894 on NCBI Build GRCm38 | |
| Downstream |
DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae) |
Dclre1b |
AI452214, Apollo, SNMIB, mSNM1B |
26.3kb | Anti-sense | Chr. 3 - 103800605 - 103809387 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15886
Genomic Map