Assay Details
| Gene Symbol: Thap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006173.1 | - | 1 - 2 | 394 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 62 | Wdr62 | 2310038K02Rik | Anti-sense | Chr. 7 - 30240138 - 30280427 on NCBI Build GRCm38 | |
| Downstream | CAP-GLY domain containing linker protein 3 | Clip3 |
1500005P14Rik, AI844915, Clipr-59 |
11.0kb | Sense | Chr. 7 - 30291688 - 30308367 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Thap8
Genomic Map