Assay Details
| Gene Symbol: 2610027K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077059.1 | 4 - 5 | 743 | 78 | ||
| GenBank mRNA | AK085522.1 | - | 4 - 5 | 567 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | breast carcinoma amplified sequence 3 | Bcas3 |
1500019F07Rik, 2610028P08Rik, AU021018, BC028339, K20D4, rudhira |
Anti-sense | Chr. 11 - 85353116 - 85826058 on NCBI Build GRCm38 | |
| Downstream | T-box 2 | Tbx2 | 36.3kb | Anti-sense | Chr. 11 - 85832615 - 85841948 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2610027K06Rik
Genomic Map