Assay Details
| Gene Symbol: Bcas3os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046194.1 | 2 - 3 | 414 | 106 | ||
| GenBank mRNA | AK020598.1 | - | 2 - 3 | 415 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5110 | Mir5110 | mmu-mir-5110 | -6.1kb | Sense | Chr. 11 - 85760616 - 85760702 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2610027K06 gene | 2610027K06Rik | 23.8kb | Sense | Chr. 11 - 85791660 - 85832388 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Bcas3os2
Genomic Map