Assay Details
| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081499.2 | NP_001074968.1 | 18 - 19 | 2992 | 85 | |
| GenBank mRNA | BC051701.1 | - | 958 | 85 | ||
| BC147581.1 | - | 18 - 19 | 2855 | 9056571 | 85 | |
| BC151057.1 | - | 18 - 19 | 2843 | 9087970 | 85 | |
More Information
Set Membership: |
|

Tbc1d8b
Genomic Map