Assay Details
| Gene Symbol: Tbc1d8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081499.2 | NP_001074968.1 | 19 - 20 | 3053 | 95 | |
| GenBank mRNA | BC051701.1 | - | 1019 | 95 | ||
| BC147581.1 | - | 19 - 20 | 2916 | 9056571 | 95 | |
| BC151057.1 | - | 19 - 20 | 2904 | 9087970 | 95 | |
More Information
Set Membership: |
|

Tbc1d8b
Genomic Map