Assay Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289653.1 | NP_001276582.1 | 11 - 12 | 1145 | 95 | |
| GenBank mRNA | AK035019.1 | - | 9 - 10 | 998 | 95 | |
| AK039790.1 | - | 1209 | 95 | |||
| AK048913.1 | - | 1416 | 95 | |||
| BC051968.1 | - | 10 - 11 | 1113 | 6401465 | 95 | |
More Information
Set Membership: |
|

Nell2
Genomic Map