Assay Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289653.1 | NP_001276582.1 | 9 - 10 | 945 | 94 | |
| GenBank mRNA | AK035019.1 | - | 7 - 8 | 798 | 94 | |
| AK039790.1 | - | 1009 | 94 | |||
| AK048913.1 | - | 1216 | 94 | |||
| BC051968.1 | - | 8 - 9 | 913 | 6401465 | 94 | |
| U59230.1 | - | 9 - 10 | 946 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nell2
Genomic Map