Assay Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026192.3 | NP_080468.1 | 2 - 3 | 318 | 83 | |
| GenBank mRNA | AK007393.1 | - | 2 - 3 | 209 | 83 | |
| AK018734.2 | - | 2 - 3 | 204 | 83 | ||
| AK035598.1 | - | 2 - 3 | 230 | 83 | ||
| AK087513.1 | - | 2 - 3 | 272 | 83 | ||
| AK220400.1 | - | 2 - 3 | 231 | 83 | ||
| AY131329.1 | - | 1 - 2 | 184 | 83 | ||
| BC054783.1 | - | 2 - 3 | 212 | 6815943 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Calcoco1
Genomic Map