Assay Details
| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021391.3 | NP_067366.1 | 2 - 3 | 258 | 101 | |
| GenBank mRNA | AF281676.1 | - | 2 - 3 | 148 | 101 | |
| BC025123.1 | - | 2 - 3 | 227 | 4019442 | 101 | |
| CA321941.1 | - | 2 - 3 | 258 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r1a
Genomic Map