Assay Details
| Gene Symbol: Ppp1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021391.3 | NP_067366.1 | 5 - 6 | 523 | 75 | |
| GenBank mRNA | AF281676.1 | - | 5 - 6 | 413 | 75 | |
| AK002813.1 | - | 1 - 2 | 157 | 75 | ||
| BC025123.1 | - | 5 - 6 | 492 | 4019442 | 75 | |
| CA321941.1 | - | 5 - 6 | 523 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r1a
Genomic Map