Assay Details
| Gene Symbol: Lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113545.1 | NP_001107017.1 | 2 - 3 | 269 | 70 | |
| GenBank mRNA | AF307845.1 | - | 2 - 3 | 223 | 70 | |
| AK049350.1 | - | 2 - 3 | 219 | 70 | ||
| BC031490.1 | - | 2 - 3 | 195 | 3498138 | 70 | |
| CJ073082.1 | - | 2 - 3 | 270 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lima1
Genomic Map