Assay Details
| Gene Symbol: Lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113545.1 | NP_001107017.1 | 5 - 6 | 871 | 69 | |
| NM_023063.4 | NP_075550.2 | 2 - 3 | 743 | 69 | ||
| GenBank mRNA | AF307844.1 | - | 2 - 3 | 666 | 69 | |
| AF307845.1 | - | 5 - 6 | 825 | 69 | ||
| AK028186.1 | - | 2 - 3 | 684 | 69 | ||
| AK031698.1 | - | 2 - 3 | 687 | 69 | ||
| AK049350.1 | - | 5 - 6 | 821 | 69 | ||
| AK085065.1 | - | 2 - 3 | 737 | 69 | ||
| AK132314.1 | - | 2 - 3 | 689 | 69 | ||
| AK167220.1 | - | 2 - 3 | 303 | 69 | ||
| BC031490.1 | - | 5 - 6 | 797 | 3498138 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lima1
Genomic Map