Assay Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031842.2 | NP_114030.2 | 11 - 12 | 1423 | 71 | |
| GenBank mRNA | AK075611.1 | - | 7 - 8 | 760 | 71 | |
| BC026783.1 | - | 11 - 12 | 1335 | 4481097 | 71 | |
| BC059921.1 | - | 11 - 12 | 1393 | 71 | ||
| BC079839.1 | - | 11 - 12 | 1369 | 6810462 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd1
Genomic Map