Assay Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031842.2 | NP_114030.2 | 5 - 6 | 695 | 67 | |
| GenBank mRNA | BC026783.1 | - | 5 - 6 | 607 | 4481097 | 67 |
| BC059921.1 | - | 5 - 6 | 665 | 67 | ||
| BC079839.1 | - | 5 - 6 | 641 | 6810462 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd1
Genomic Map