Assay Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031842.2 | NP_114030.2 | 9 - 10 | 1170 | 74 | |
| GenBank mRNA | AK075611.1 | - | 5 - 6 | 507 | 74 | |
| BC026783.1 | - | 9 - 10 | 1082 | 4481097 | 74 | |
| BC059921.1 | - | 9 - 10 | 1140 | 74 | ||
| BC079839.1 | - | 9 - 10 | 1116 | 6810462 | 74 | |
| M25773.1 | - | 9 - 10 | 1126 | 74 | ||
| U66620.1 | - | 9 - 10 | 1126 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd1
Genomic Map