Assay Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003873.1 | - | 1 - 2 | 121 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003873.1 transcripts.
Set Membership: |
|

Rnasel
Genomic Map