Assay Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011882.2 | NP_036012.1 | 2 - 3 | 383 | 66 | |
| GenBank mRNA | AK053125.1 | - | 1 - 2 | 109 | 66 | |
| AK140815.1 | - | 2 - 3 | 384 | 66 | ||
| BC003873.1 | - | 2 - 3 | 192 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003873.1, NM_011882.2 transcripts.
Set Membership: |
|

Rnasel
Genomic Map