Assay Details
| Gene Symbol: Rgs16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011267.3 | NP_035397.2 | 2 - 3 | 271 | 64 | |
| GenBank mRNA | AK079543.1 | - | 2 - 3 | 272 | 64 | |
| AK154324.1 | - | 2 - 3 | 270 | 64 | ||
| BC010980.1 | - | 2 - 3 | 264 | 4020813 | 64 | |
| BC049968.1 | - | 2 - 3 | 259 | 4021479 | 64 | |
| BC055459.1 | - | 2 - 3 | 275 | 4194934 | 64 | |
| U67189.1 | - | 2 - 3 | 261 | 64 | ||
| U72881.1 | - | 2 - 3 | 226 | 64 | ||
| U94828.1 | - | 2 - 3 | 264 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs16
Genomic Map