Assay Details
| Gene Symbol: Rgs16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011267.3 | NP_035397.2 | 3 - 4 | 337 | 65 | |
| GenBank mRNA | AK079543.1 | - | 3 - 4 | 338 | 65 | |
| AK154324.1 | - | 3 - 4 | 336 | 65 | ||
| AK193821.1 | - | 2 - 3 | 159 | 65 | ||
| BC010980.1 | - | 3 - 4 | 330 | 4020813 | 65 | |
| BC049968.1 | - | 3 - 4 | 325 | 4021479 | 65 | |
| U67189.1 | - | 3 - 4 | 327 | 65 | ||
| U72881.1 | - | 3 - 4 | 292 | 65 | ||
| U94828.1 | - | 3 - 4 | 330 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs16
Genomic Map