Assay Details
| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152804.2 | NP_690017.2 | 2 - 3 | 506 | 67 | |
| GenBank mRNA | AK133962.1 | - | 2 - 3 | 504 | 67 | |
| AK143452.1 | - | 1 - 2 | 1081 | 67 | ||
| AK164365.1 | - | 2 - 3 | 505 | 67 | ||
| BC034513.1 | - | 2 - 3 | 488 | 3156743 | 67 | |
| BC035451.1 | - | 2 - 3 | 488 | 67 | ||
| BM947259.1 | - | 2 - 3 | 448 | 67 | ||
| M96163.1 | - | 2 - 3 | 472 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk2
Genomic Map