Assay Details
| Gene Symbol: Plk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152804.2 | NP_690017.2 | 7 - 8 | 1137 | 78 | |
| GenBank mRNA | AK133962.1 | - | 7 - 8 | 1136 | 78 | |
| AK164365.1 | - | 7 - 8 | 1136 | 78 | ||
| BC034513.1 | - | 7 - 8 | 1119 | 3156743 | 78 | |
| BC035451.1 | - | 7 - 8 | 1119 | 78 | ||
| M96163.1 | - | 7 - 8 | 1103 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk2
Genomic Map