Assay Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286743.1 | NP_001273672.1 | 5 - 6 | 930 | 65 | |
| NM_001286744.1 | NP_001273673.1 | 5 - 6 | 836 | 65 | ||
| NM_011861.3 | NP_035991.1 | 5 - 6 | 895 | 65 | ||
| NM_178365.4 | NP_848142.1 | 5 - 6 | 913 | 65 | ||
| GenBank mRNA | AK043967.1 | - | 5 - 6 | 899 | 65 | |
| AK158686.1 | - | 5 - 6 | 933 | 65 | ||
| BC014698.1 | - | 5 - 6 | 784 | 4527708 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pacsin1
Genomic Map