Assay Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281491.1 | NP_001268420.1 | 10 - 11 | 1677 | 65 | |
| GenBank mRNA | CB525601.1 | - | 1 - 2 | 115 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syngap1
Genomic Map