Assay Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281491.1 | NP_001268420.1 | 14 - 15 | 2330 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syngap1| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281491.1 | NP_001268420.1 | 14 - 15 | 2330 | 65 | |
Set Membership: |
|