Assay Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281491.1 | NP_001268420.1 | 15 - 16 | 3402 | 73 | |
| GenBank mRNA | BM939748.1 | - | 1 - 2 | 416 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syngap1
Genomic Map