Assay Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301373.1 | NP_001288302.1 | 2 - 3 | 638 | 100 | |
| NM_001301375.1 | NP_001288304.1 | 2 - 3 | 638 | 100 | ||
| NM_008046.3 | NP_032072.1 | 2 - 3 | 638 | 100 | ||
| GenBank mRNA | AK079916.1 | - | 2 - 3 | 639 | 100 | |
| AK083556.1 | - | 2 - 3 | 417 | 100 | ||
| BC144926.1 | - | 2 - 3 | 462 | 9053474 | 100 | |
| BC145945.1 | - | 2 - 3 | 462 | 40131248 | 100 | |
| X83377.1 | - | 2 - 3 | 267 | 100 | ||
| Z29532.1 | - | 2 - 3 | 273 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fst
Genomic Map