Assay Details
| Gene Symbol: Pim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008842.3 | NP_032868.2 | 1 - 2 | 181 | 70 | |
| GenBank mRNA | AK158445.1 | - | 1 - 2 | 433 | 70 | |
| BC042885.1 | - | 1 - 2 | 414 | 4507947 | 70 | |
| BC053019.1 | - | 1 - 2 | 181 | 5694941 | 70 | |
| BC055316.1 | - | 1 - 2 | 181 | 5694941 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pim1
Genomic Map