Assay Details
| Gene Symbol: Taf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026836.2 | NP_081112.1 | 4 - 5 | 562 | 81 | |
| GenBank mRNA | AK004168.1 | - | 3 - 4 | 209 | 81 | |
| AK050689.1 | - | 4 - 5 | 563 | 81 | ||
| AK133851.1 | - | 5 - 6 | 1546 | 81 | ||
| BC005603.1 | - | 4 - 5 | 541 | 3483315 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
positive regulation by host of viral transcription
RNA polymerase II transcriptional preinitiation complex assembly
regulation of nucleic acid-templated transcription
DNA binding
transcription coactivator activity
transcription factor binding
vitamin D receptor binding
thyroid hormone receptor binding
protein heterodimerization activity
protein N-terminus binding
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
positive regulation by host of viral transcription
RNA polymerase II transcriptional preinitiation complex assembly
regulation of nucleic acid-templated transcription
DNA binding
transcription coactivator activity
transcription factor binding
vitamin D receptor binding
thyroid hormone receptor binding
protein heterodimerization activity
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119183 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119191 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119192 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092073 | NM_026836.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092073 | AK050689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092073 | AK133851.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12199 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169388 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092073 | NM_026836.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092073 | AK050689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092073 | AK133851.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P230628 | BC005603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P230628 | NM_026836.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P230628 | AK004168.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P230628 | AK050689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P230628 | AK133851.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P410127 | BC005603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P410127 | NM_026836.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P410127 | AK004168.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P410127 | AK050689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P410127 | AK133851.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31119185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169388 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08749285 | - | - |
| Whole Mouse Genome G4122A | A_52_P230628 | BC005603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P230628 | NM_026836.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P230628 | AK004168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P230628 | AK050689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P230628 | AK133851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410127 | BC005603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410127 | NM_026836.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410127 | AK004168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410127 | AK050689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410127 | AK133851.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132145 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169388 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13305204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13305206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13305203 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13305195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13203896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13305201 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119191 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31119183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31119187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13429117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13305203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13537971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13305206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31119142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449400 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451995_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449394 | - | - |
| Mouse Genome 430 2.0 Array | 1446161_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446161_at | - | - |
| Mouse Genome 430 2.0 Array | 1451995_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451995_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68776_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68776_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700016082.mm.1 | NM_026836.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016082.mm.1 | NM_026836.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016082.mm.1 | TC1700001747.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016084.mm.1 | NM_026836.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016084.mm.1 | NM_026836.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016084.mm.1 | TC1700001747.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016083.mm.1 | NM_026836.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016083.mm.1 | NM_026836.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016083.mm.1 | TC1700001747.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451995_PM_at | - | - |
| Mouse Expression Set 430 | 1451995_at | - | - |
| Murine Genome U74v2 Set | 96691_at | - | - |
| Murine Genome U74v2 Set | 96691_PM_at | - | - |
| Murine Genome U74v2 Set | 170787_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170704_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170704_i_at | - | - |
| Murine Genome U74v2 Set | 170787_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451995_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451995_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451995_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451995_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 761588 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223605 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223605 | - | - |
| mouse-6 v1.1 (Illumina) | scl068776.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068776.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2946281 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2946281 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068776.1_229-S | - | - |

Taf11
Genomic Map