Assay Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033218.1 | NP_150087.1 | 10 - 11 | 2140 | 85 | |
| GenBank mRNA | AF374267.1 | - | 10 - 11 | 1993 | 85 | |
| AK155857.1 | - | 11 - 12 | 2088 | 85 | ||
| AK172219.1 | - | 10 - 11 | 2147 | 85 | ||
| BQ963055.1 | - | 1 - 2 | 195 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srebf2
Genomic Map