Assay Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033218.1 | NP_150087.1 | 17 - 18 | 3197 | 65 | |
| GenBank mRNA | AF374267.1 | - | 17 - 18 | 3050 | 65 | |
| AK155857.1 | - | 18 - 19 | 3145 | 65 | ||
| AK172219.1 | - | 17 - 18 | 3206 | 65 | ||
| CB228109.2 | - | 2 - 3 | 397 | 65 | ||
More Information
Set Membership: |
|

Srebf2
Genomic Map