Assay Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033218.1 | NP_150087.1 | 2 - 3 | 637 | 73 | |
| GenBank mRNA | AF374267.1 | - | 2 - 3 | 490 | 73 | |
| AK053802.1 | - | 2 - 3 | 656 | 73 | ||
| AK155857.1 | - | 3 - 4 | 585 | 73 | ||
| AK172219.1 | - | 2 - 3 | 642 | 73 | ||
| BC014730.1 | - | 2 - 3 | 627 | 73 | ||
| BC023333.1 | - | 1 - 2 | 306 | 73 | ||
| BC026556.1 | - | 2 - 3 | 629 | 73 | ||
| BC038284.1 | - | 2 - 3 | 628 | 73 | ||
| BC048071.1 | - | 2 - 3 | 642 | 73 | ||
| BC069989.1 | - | 2 - 3 | 646 | 73 | ||
| BQ965109.1 | - | 2 - 3 | 644 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srebf2
Genomic Map