Assay Details
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033218.1 | NP_150087.1 | 4 - 5 | 966 | 106 | |
GenBank mRNA | AF289715.2 | - | 2 - 3 | 293 | 106 | |
AF374267.1 | - | 4 - 5 | 819 | 106 | ||
AK155857.1 | - | 5 - 6 | 914 | 106 | ||
AK172219.1 | - | 4 - 5 | 972 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
