Assay Details
| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253702.1 | NP_001240631.1 | 6 - 7 | 1307 | 74 | |
| NM_153091.3 | NP_694731.1 | 6 - 7 | 1307 | 74 | ||
| NR_045578.1 | 6 - 7 | 1573 | 74 | |||
| GenBank mRNA | AB084518.1 | - | 6 - 7 | 721 | 74 | |
| AK033865.1 | - | 6 - 7 | 739 | 74 | ||
| AK148099.1 | - | 6 - 7 | 1009 | 74 | ||
| AK172102.1 | - | 6 - 7 | 1308 | 74 | ||
| AK172218.1 | - | 6 - 7 | 1308 | 74 | ||
| BC024473.1 | - | 6 - 7 | 698 | 74 | ||
| BC058781.1 | - | 6 - 7 | 732 | 6311899 | 74 | |
| BC107394.2 | - | 6 - 7 | 732 | 40058131 | 74 | |
| BC145950.1 | - | 6 - 7 | 799 | 40131253 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

St7l
Genomic Map