Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_001253702.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_001253702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_001253703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_001253703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_153091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NM_153091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NR_045578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
NR_045578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008774.mm.1 |
TC0300001083.mm.1 |
MTA TC |